Assay Details
| Gene Symbol: Pgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033537.2 | NP_001028709.2 | 5 - 6 | 648 | 77 | |
| GenBank mRNA | AK052486.1 | - | 5 - 6 | 565 | 77 | |
| AK078974.1 | - | 5 - 6 | 574 | 77 | ||
| AK170817.1 | - | 5 - 6 | 563 | 77 | ||
| BC030368.1 | - | 3 - 4 | 268 | 77 | ||
| BC035945.1 | - | 5 - 6 | 571 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pgap3
Genomic Map