Assay Details
| Gene Symbol: Pgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033537.2 | NP_001028709.2 | 6 - 7 | 787 | 62 | |
| GenBank mRNA | AK052486.1 | - | 6 - 7 | 704 | 62 | |
| AK078974.1 | - | 6 - 7 | 713 | 62 | ||
| AK170817.1 | - | 6 - 7 | 702 | 62 | ||
| BC030368.1 | - | 4 - 5 | 407 | 62 | ||
| BC035945.1 | - | 6 - 7 | 710 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pgap3
Genomic Map