Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
NM_001113374.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
NM_001113374.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
NM_001113375.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
NM_001113375.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
NM_013826.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
NM_013826.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008283.mm.1 |
TC1300001318.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008284.mm.1 |
NM_001113374.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008284.mm.1 |
NM_001113374.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008284.mm.1 |
NM_013826.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008284.mm.1 |
NM_013826.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008284.mm.1 |
TC1300001318.mm.1 |
MTA TC |