Assay Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113374.1 | NP_001106845.1 | 4 - 5 | 512 | 68 | |
| NM_013826.3 | NP_038854.2 | 5 - 6 | 634 | 68 | ||
| GenBank mRNA | AK002719.1 | - | 4 - 5 | 480 | 68 | |
| AK010112.1 | - | 5 - 6 | 614 | 68 | ||
| BC004631.1 | - | 4 - 5 | 483 | 68 | ||
| BC024371.1 | - | 5 - 6 | 594 | 4983231 | 68 | |
More Information
Set Membership: |
|

Mocs2
Genomic Map