Assay Details
| Gene Symbol: Il31ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030512.1 | - | 1 - 2 | 211 | 64 | |
| BC145083.1 | - | 12 - 13 | 1651 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030512.1 transcripts.
Set Membership: |
|

Il31ra
Genomic Map