Assay Details
| Gene Symbol: Il31ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139299.2 | NP_647460.2 | 2 - 3 | 185 | 98 | |
| GenBank mRNA | AB083111.1 | - | 2 - 3 | 185 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB083111.1, NM_139299.2 transcripts.
Set Membership: |
|

Il31ra
Genomic Map