Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 10 - 11 | 1187 | 142 | |
| GenBank mRNA | AK044487.1 | - | 10 - 11 | 1219 | 142 | |
| AK087793.1 | - | 10 - 11 | 1220 | 142 | ||
| BC029746.1 | - | 10 - 11 | 1187 | 3494321 | 142 | |
| BC031900.1 | - | 10 - 11 | 1123 | 5364208 | 142 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ipo11
Genomic Map