Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 11 - 12 | 1337 | 77 | |
| GenBank mRNA | AK044487.1 | - | 11 - 12 | 1369 | 77 | |
| AK087793.1 | - | 11 - 12 | 1370 | 77 | ||
| BC029746.1 | - | 11 - 12 | 1337 | 3494321 | 77 | |
| BC031900.1 | - | 11 - 12 | 1273 | 5364208 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo11
Genomic Map