Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 15 - 16 | 1618 | 131 | |
| GenBank mRNA | AK044487.1 | - | 15 - 16 | 1650 | 131 | |
| AK087793.1 | - | 15 - 16 | 1651 | 131 | ||
| BC029746.1 | - | 15 - 16 | 1618 | 3494321 | 131 | |
| BC031900.1 | - | 15 - 16 | 1554 | 5364208 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo11
Genomic Map