Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 16 - 17 | 1748 | 158 | |
| GenBank mRNA | AK044487.1 | - | 16 - 17 | 1780 | 158 | |
| AK087793.1 | - | 16 - 17 | 1781 | 158 | ||
| BC029746.1 | - | 16 - 17 | 1748 | 3494321 | 158 | |
| BC031900.1 | - | 16 - 17 | 1684 | 5364208 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo11
Genomic Map