Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 18 - 19 | 1828 | 114 | |
| GenBank mRNA | AK044487.1 | - | 18 - 19 | 1860 | 114 | |
| AK087793.1 | - | 18 - 19 | 1861 | 114 | ||
| BC029746.1 | - | 18 - 19 | 1828 | 3494321 | 114 | |
| BC031900.1 | - | 18 - 19 | 1764 | 5364208 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo11
Genomic Map