Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 21 - 22 | 2171 | 101 | |
| GenBank mRNA | AK044487.1 | - | 21 - 22 | 2203 | 101 | |
| AK087793.1 | - | 21 - 22 | 2204 | 101 | ||
| BC029746.1 | - | 21 - 22 | 2171 | 3494321 | 101 | |
| BC031900.1 | - | 21 - 22 | 2107 | 5364208 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo11
Genomic Map