Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 22 - 23 | 2248 | 144 | |
| GenBank mRNA | AK087793.1 | - | 22 - 23 | 2281 | 144 | |
More Information
Set Membership: |
|

Ipo11
Genomic Map