Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159525.1 | NP_001152997.1 | 1 - 2 | 95 | 82 | |
| GenBank mRNA | AK028449.1 | - | 1 - 2 | 89 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pex19
Genomic Map