Assay Details
| Gene Symbol: Dcaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177267.4 | NP_796241.3 | 6 - 7 | 1079 | 90 | |
| GenBank mRNA | AK122561.1 | - | 6 - 7 | 916 | 90 | |
| AK137579.1 | - | 6 - 7 | 1079 | 90 | ||
| AK165096.1 | - | 6 - 7 | 1063 | 90 | ||
| BC042567.1 | - | 6 - 7 | 753 | 90 | ||
| BC046813.1 | - | 6 - 7 | 1017 | 5702325 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf5
Genomic Map