Assay Details
| Gene Symbol: Tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145968.2 | NP_666080.1 | 2 - 3 | 183 | 112 | |
| GenBank mRNA | BC030886.1 | - | 2 - 3 | 163 | 4483459 | 112 |
| BC039780.1 | - | 2 - 3 | 174 | 4483459 | 112 | |
| BC150728.1 | - | 2 - 3 | 128 | 9087639 | 112 | |
| BY117429.1 | - | 2 - 3 | 185 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC030886.1, BC039780.1, BC150728.1, NM_145968.2 transcripts.
Set Membership: |
|

Tagap
Genomic Map