Assay Details
| Gene Symbol: Tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145968.2 | NP_666080.1 | 6 - 7 | 632 | 102 | |
| GenBank mRNA | BC030886.1 | - | 6 - 7 | 612 | 4483459 | 102 |
| BC039780.1 | - | 6 - 7 | 623 | 4483459 | 102 | |
| BC150728.1 | - | 6 - 7 | 577 | 9087639 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tagap
Genomic Map