Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 12 - 13 | 1948 | 96 | |
| GenBank mRNA | AK088000.1 | - | 12 - 13 | 1909 | 96 | |
| AK137001.1 | - | 12 - 13 | 1907 | 96 | ||
| AK150523.1 | - | 5 - 6 | 853 | 96 | ||
| AK159279.1 | - | 12 - 13 | 1905 | 96 | ||
| AK159306.1 | - | 12 - 13 | 1903 | 96 | ||
| BC010319.1 | - | 10 - 11 | 1533 | 96 | ||
| BC021496.1 | - | 12 - 13 | 1898 | 5346159 | 96 | |
| BC023794.1 | - | 13 - 14 | 1843 | 5351943 | 96 | |
| BC023964.1 | - | 12 - 13 | 1888 | 96 | ||
| BC025037.1 | - | 5 - 6 | 748 | 96 | ||
| BC025618.1 | - | 12 - 13 | 1907 | 5320976 | 96 | |
| BC025627.1 | - | 12 - 13 | 1898 | 5320513 | 96 | |
| BC025811.1 | - | 12 - 13 | 1885 | 5096872 | 96 | |
| BC032187.1 | - | 12 - 13 | 1948 | 5321268 | 96 | |
| BC033435.1 | - | 12 - 13 | 1844 | 4950746 | 96 | |
| BC033471.1 | - | 12 - 13 | 1888 | 4954119 | 96 | |
| BC034177.1 | - | 11 - 12 | 1653 | 96 | ||
| BC035352.1 | - | 11 - 12 | 1618 | 96 | ||
| BC042435.1 | - | 12 - 13 | 1898 | 4951288 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map