Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 14 - 15 | 2258 | 65 | |
| GenBank mRNA | AK088000.1 | - | 14 - 15 | 2219 | 65 | |
| AK137001.1 | - | 14 - 15 | 2217 | 65 | ||
| AK148989.1 | - | 1 - 2 | 116 | 65 | ||
| AK150523.1 | - | 7 - 8 | 1163 | 65 | ||
| AK159279.1 | - | 14 - 15 | 2215 | 65 | ||
| AK159306.1 | - | 14 - 15 | 2213 | 65 | ||
| BC010319.1 | - | 12 - 13 | 1843 | 65 | ||
| BC021496.1 | - | 14 - 15 | 2208 | 5346159 | 65 | |
| BC023794.1 | - | 15 - 16 | 2153 | 5351943 | 65 | |
| BC023964.1 | - | 14 - 15 | 2199 | 65 | ||
| BC025037.1 | - | 7 - 8 | 1058 | 65 | ||
| BC025618.1 | - | 14 - 15 | 2217 | 5320976 | 65 | |
| BC025627.1 | - | 14 - 15 | 2208 | 5320513 | 65 | |
| BC025811.1 | - | 14 - 15 | 2195 | 5096872 | 65 | |
| BC030326.1 | - | 3 - 4 | 555 | 65 | ||
| BC031176.1 | - | 1 - 2 | 116 | 65 | ||
| BC032187.1 | - | 14 - 15 | 2258 | 5321268 | 65 | |
| BC033435.1 | - | 14 - 15 | 2154 | 4950746 | 65 | |
| BC033471.1 | - | 14 - 15 | 2198 | 4954119 | 65 | |
| BC034177.1 | - | 13 - 14 | 1963 | 65 | ||
| BC035352.1 | - | 13 - 14 | 1928 | 65 | ||
| BC042435.1 | - | 14 - 15 | 2208 | 4951288 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map