Assay Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 21 - 22 | 3240 | 90 | |
| GenBank mRNA | AK088000.1 | - | 21 - 22 | 3201 | 90 | |
| AK137001.1 | - | 21 - 22 | 3200 | 90 | ||
| AK148989.1 | - | 8 - 9 | 1098 | 90 | ||
| AK150523.1 | - | 14 - 15 | 2145 | 90 | ||
| AK159279.1 | - | 21 - 22 | 3197 | 90 | ||
| AK159306.1 | - | 21 - 22 | 3195 | 90 | ||
| BC010319.1 | - | 19 - 20 | 2825 | 90 | ||
| BC021496.1 | - | 21 - 22 | 3190 | 5346159 | 90 | |
| BC023175.1 | - | 5 - 6 | 534 | 90 | ||
| BC023794.1 | - | 22 - 23 | 3135 | 5351943 | 90 | |
| BC023867.1 | - | 4 - 5 | 536 | 90 | ||
| BC023964.1 | - | 21 - 22 | 3181 | 90 | ||
| BC025037.1 | - | 14 - 15 | 2040 | 90 | ||
| BC025618.1 | - | 21 - 22 | 3199 | 5320976 | 90 | |
| BC025627.1 | - | 21 - 22 | 3190 | 5320513 | 90 | |
| BC025811.1 | - | 21 - 22 | 3177 | 5096872 | 90 | |
| BC030326.1 | - | 10 - 11 | 1537 | 90 | ||
| BC031176.1 | - | 8 - 9 | 1098 | 90 | ||
| BC031389.1 | - | 5 - 6 | 659 | 90 | ||
| BC032187.1 | - | 21 - 22 | 3240 | 5321268 | 90 | |
| BC033435.1 | - | 21 - 22 | 3136 | 4950746 | 90 | |
| BC033471.1 | - | 21 - 22 | 3180 | 4954119 | 90 | |
| BC034177.1 | - | 20 - 21 | 2945 | 90 | ||
| BC035352.1 | - | 20 - 21 | 2910 | 90 | ||
| BC042435.1 | - | 21 - 22 | 3190 | 4951288 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a1
Genomic Map