Assay Details
| Gene Symbol: Aaed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025370.2 | NP_079646.1 | 5 - 6 | 577 | 68 | |
| GenBank mRNA | AK003782.1 | - | 5 - 6 | 578 | 68 | |
| AK006589.1 | - | 5 - 6 | 432 | 68 | ||
| AK145073.1 | - | 575 | 68 | |||
| AK148379.1 | - | 578 | 68 | |||
| AK168581.1 | - | 4 - 5 | 389 | 68 | ||
| AK168701.1 | - | 456 | 68 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098293 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009351 | AK168581.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009351 | AK168701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009351 | AK145073.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009351 | AK145867.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009351 | AK006589.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151069 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117909 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129238 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016193 | AK168701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016193 | AK145867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009351 | AK168581.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009351 | AK168701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009351 | AK145073.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009351 | AK145867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009351 | AK006589.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29098283 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02709656 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117909 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02709655 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07900882 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129238 | - | - |
| Whole Mouse Genome G4122A | A_51_P361049 | AK148379.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361049 | AK168581.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361049 | AK168701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361049 | AK145073.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361049 | AK145867.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108423 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02709655 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117909 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10929178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10875919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10929174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098293 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29098288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29098188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10455014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10929178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11093979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11083158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10882289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11000739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10839329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11063275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10682507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10498793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10939497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10690842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10898392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10929183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11057252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29098224 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10410116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10410122 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421018_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434441_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10410115 | - | - |
| Mouse Genome 430 2.0 Array | 1434441_at | - | - |
| Mouse Genome 430 2.0 Array | 1421018_at | - | - |
| Mouse Genome 430 2.0 Array | 1434441_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421018_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66129_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66129_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300014214.mm.1 | NM_025370.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014214.mm.1 | NM_025370.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014214.mm.1 | TC1300002201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300014215.mm.1 | NM_025370.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014215.mm.1 | NM_025370.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300014215.mm.1 | TC1300002201.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421018_PM_at | - | - |
| Mouse Expression Set 430 | 1421018_at | - | - |
| Mouse Expression Set 430 | 1434441_PM_at | - | - |
| Mouse Expression Set 430 | 1434441_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421018_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421018_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434441_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434441_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 70161 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2647669 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2674655 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2674655 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2647669 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003641.1_23-S | - | - |
| mouse-6 v1.1 (Illumina) | scl066129.2_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066129.2_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003641.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810671 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2674655 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2647669 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2647669 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2674655 | - | - |

Aaed1
Genomic Map