Assay Details
| Gene Symbol: Ctsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009984.3 | NP_034114.1 | 1 - 2 | 70 | 71 | |
| GenBank mRNA | AK049466.1 | - | 1 - 2 | 71 | 71 | |
| AK145051.1 | - | 1 - 2 | 71 | 71 | ||
| AK147142.1 | - | 1 - 2 | 72 | 71 | ||
| AK150452.1 | - | 1 - 2 | 71 | 71 | ||
| AK151169.1 | - | 1 - 2 | 71 | 71 | ||
| AK153076.1 | - | 1 - 2 | 71 | 71 | ||
| AK153417.1 | - | 1 - 2 | 71 | 71 | ||
| AK159511.1 | - | 1 - 2 | 71 | 71 | ||
| AK160100.1 | - | 1 - 2 | 71 | 71 | ||
| AK161538.1 | - | 1 - 2 | 71 | 71 | ||
| AK165397.1 | - | 1 - 2 | 71 | 71 | ||
| AK169394.1 | - | 1 - 2 | 71 | 71 | ||
| BC068163.1 | - | 1 - 2 | 56 | 30359160 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049466.1, AK145051.1, AK147142.1, AK150452.1, AK151169.1, AK153076.1, AK153417.1, AK159511.1, AK160100.1, AK161538.1, AK165397.1, AK169394.1, BC068163.1, NM_009984.3 transcripts.
Set Membership: |
|

Ctsl
Genomic Map