Assay Details
| Gene Symbol: 1700006H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045900.2 | 1 - 2 | 183 | 100 | ||
| GenBank mRNA | AK005677.1 | - | 1 - 2 | 184 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 32090 |
Gm32090 | -23.0kb | Anti-sense | Chr. 13 - 107605292 - 107664374 on NCBI Build GRCm38 | |
| Downstream | zinc finger SWIM-type containing 6 | Zswim6 |
2900036G02Rik, mKIAA1577 |
34.5kb | Anti-sense | Chr. 13 - 107724617 - 107890064 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700006H21Rik
Genomic Map