Assay Details
| Gene Symbol: 1700006H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045900.2 | 2 - 3 | 343 | 85 | ||
| GenBank mRNA | AK005677.1 | - | 2 - 3 | 344 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 32090 |
Gm32090 | -25.6kb | Anti-sense | Chr. 13 - 107605292 - 107664374 on NCBI Build GRCm38 | |
| Downstream | zinc finger SWIM-type containing 6 | Zswim6 |
2900036G02Rik, mKIAA1577 |
32.4kb | Anti-sense | Chr. 13 - 107724617 - 107890064 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172389 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156547 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156547 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133951 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10904964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10863892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11043766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10692732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10977357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10561640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10689813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11043765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10457652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10813634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11113095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10689815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10736008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10980364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10692711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10527891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10436736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29306689 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1300007624.mm.1 | NR_045900.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300007624.mm.1 | TC1300001198.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 772024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104280161 | - | - |

1700006H21Rik
Genomic Map