Assay Details
| Gene Symbol: Numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136075.2 | NP_001129547.1 | 9 - 10 | 1258 | 69 | |
| NM_001272055.1 | NP_001258984.1 | 8 - 9 | 1225 | 69 | ||
| NR_073564.1 | 12 - 13 | 1471 | 69 | |||
| GenBank mRNA | AF169191.1 | - | 8 - 9 | 1210 | 69 | |
| AF169192.1 | - | 9 - 10 | 1243 | 69 | ||
| BC050108.1 | - | 12 - 13 | 1381 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Numb
Genomic Map