Assay Details
| Gene Symbol: Znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC089554.1 | - | 2 - 3 | 122 | 30300471 | 62 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC089554.1 transcripts.
Set Membership: |
|

Znhit3
Genomic Map