Assay Details
| Gene Symbol: Car10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028296.3 | NP_082572.1 | 6 - 7 | 1573 | 101 | |
| GenBank mRNA | AB080741.1 | - | 7 - 8 | 1529 | 101 | |
| AK132222.1 | - | 6 - 7 | 1414 | 101 | ||
| BC017606.1 | - | 6 - 7 | 765 | 4507552 | 101 | |
| BC066111.1 | - | 7 - 8 | 1457 | 30360743 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car10
Genomic Map