Assay Details
| Gene Symbol: Car10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028296.3 | NP_082572.1 | 7 - 8 | 1725 | 112 | |
| GenBank mRNA | AB080741.1 | - | 8 - 9 | 1681 | 112 | |
| AK132222.1 | - | 7 - 8 | 1566 | 112 | ||
| BC017606.1 | - | 7 - 8 | 917 | 4507552 | 112 | |
| BC066111.1 | - | 8 - 9 | 1609 | 30360743 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car10
Genomic Map