Assay Details
| Gene Symbol: Tom1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028011.2 | NP_082287.1 | 1 - 2 | 89 | 69 | |
| GenBank mRNA | AF395837.1 | - | 1 - 2 | 70 | 69 | |
| AJ505007.1 | - | 1 - 2 | 67 | 69 | ||
| AK009826.1 | - | 1 - 2 | 69 | 69 | ||
| AK033712.1 | - | 1 - 2 | 91 | 69 | ||
| AK083630.1 | - | 68 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033712.1, NM_028011.2 transcripts.
Set Membership: |
|

Tom1l1
Genomic Map