Assay Details
| Gene Symbol: Abi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163464.1 | NP_001156936.1 | 1 - 2 | 70 | 70 | |
| GenBank mRNA | AK037144.1 | - | 1 - 2 | 71 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037144.1, NM_001163464.1 transcripts.
Set Membership: |
|

Abi3
Genomic Map