Assay Details
| Gene Symbol: Eml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162996.1 | NP_001156468.1 | 13 - 14 | 1356 | 70 | |
| NM_028153.1 | NP_082429.1 | 13 - 14 | 1413 | 70 | ||
| GenBank mRNA | AK005562.1 | - | 13 - 14 | 1379 | 70 | |
| BC055476.1 | - | 13 - 14 | 1356 | 4218133 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eml2
Genomic Map