Assay Details
| Gene Symbol: Eml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162996.1 | NP_001156468.1 | 1 - 2 | 54 | 64 | |
| NM_028153.1 | NP_082429.1 | 1 - 2 | 54 | 64 | ||
| GenBank mRNA | BC055476.1 | - | 1 - 2 | 54 | 4218133 | 64 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC055476.1, NM_001162996.1, NM_028153.1 transcripts.
Set Membership: |
|

Eml2
Genomic Map