Assay Details
| Gene Symbol: Vsx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301427.1 | NP_001288356.1 | 3 - 4 | 691 | 62 | |
| NM_007701.3 | NP_031727.1 | 3 - 4 | 691 | 62 | ||
| GenBank mRNA | AK143461.1 | - | 3 - 4 | 691 | 62 | |
| BC031869.1 | - | 3 - 4 | 667 | 4480964 | 62 | |
| BC051245.1 | - | 3 - 4 | 656 | 6494544 | 62 | |
| BC058806.1 | - | 3 - 4 | 679 | 6492679 | 62 | |
| L34808.1 | - | 3 - 4 | 576 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vsx2
Genomic Map