Assay Details
| Gene Symbol: Med6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027213.2 | NP_081489.2 | 2 - 3 | 181 | 100 | |
| GenBank mRNA | AK089101.1 | - | 2 - 3 | 230 | 100 | |
| AK171784.1 | - | 2 - 3 | 197 | 100 | ||
| BC013096.1 | - | 2 - 3 | 181 | 3710612 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC013096.1, NM_027213.2 transcripts.
Set Membership: |
|

Med6
Genomic Map