Assay Details
| Gene Symbol: Med6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027213.2 | NP_081489.2 | 4 - 5 | 354 | 69 | |
| GenBank mRNA | AK005232.1 | - | 4 - 5 | 399 | 69 | |
| AK089101.1 | - | 4 - 5 | 403 | 69 | ||
| AK171784.1 | - | 4 - 5 | 370 | 69 | ||
| BC013096.1 | - | 4 - 5 | 354 | 3710612 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med6
Genomic Map