Assay Details
| Gene Symbol: Med6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027213.2 | NP_081489.2 | 5 - 6 | 462 | 107 | |
| GenBank mRNA | AK005232.1 | - | 5 - 6 | 507 | 107 | |
| AK171784.1 | - | 5 - 6 | 478 | 107 | ||
| BC013096.1 | - | 5 - 6 | 462 | 3710612 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med6
Genomic Map