Assay Details
| Gene Symbol: 1810021B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040417.1 | 1 - 2 | 238 | 81 | ||
| GenBank mRNA | AK007571.1 | - | 1 - 2 | 239 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pannexin 2 | Panx2 | -502.0 | Anti-sense | Chr. 15 - 89059726 - 89073571 on NCBI Build GRCm38 | |
| Downstream | TraB domain containing | Trabd |
5730502D15Rik, AL023039 |
196.0 | Anti-sense | Chr. 15 - 89076050 - 89087078 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1810021B22Rik
Genomic Map