Assay Details
| Gene Symbol: Cdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042657.1 | - | 2 - 3 | 302 | 55 | |
| AK140452.1 | - | 3 - 4 | 463 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042657.1 transcripts.
Set Membership: |
|

Cdh7
Genomic Map