Assay Details
| Gene Symbol: Cdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY267034.1 | - | 5 - 6 | 1183 | 81 | |
| BC141119.1 | - | 5 - 6 | 1159 | 9055731 | 81 | |
| BC145423.1 | - | 6 - 7 | 1277 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh7
Genomic Map