Assay Details
| Gene Symbol: Krt84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008474.2 | NP_032500.2 | 6 - 7 | 1292 | 96 | |
| GenBank mRNA | AY028607.1 | - | 6 - 7 | 1249 | 96 | |
| BC114971.1 | - | 6 - 7 | 1245 | 40039508 | 96 | |
| X65505.1 | - | 3 - 4 | 312 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt84
Genomic Map