Assay Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139140.1 | NP_631879.1 | 3 - 4 | 422 | 88 | |
| GenBank mRNA | AB079132.1 | - | 3 - 4 | 422 | 88 | |
| AK048720.1 | - | 3 - 4 | 464 | 88 | ||
| AK135838.1 | - | 3 - 4 | 480 | 88 | ||
| AK159335.1 | - | 3 - 4 | 516 | 88 | ||
| AK167788.1 | - | 2 - 3 | 99 | 88 | ||
| AK171266.1 | - | 2 - 3 | 177 | 88 | ||
| BC023432.1 | - | 1 - 2 | 72 | 5029821 | 88 | |
| BC072579.1 | - | 3 - 4 | 689 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023432.1 transcripts.
Set Membership: |
|

Spats2
Genomic Map