Assay Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139140.1 | NP_631879.1 | 7 - 8 | 1021 | 67 | |
| GenBank mRNA | AB079132.1 | - | 7 - 8 | 1021 | 67 | |
| AK048720.1 | - | 7 - 8 | 1063 | 67 | ||
| AK135838.1 | - | 7 - 8 | 1079 | 67 | ||
| AK159335.1 | - | 7 - 8 | 1116 | 67 | ||
| AK167788.1 | - | 6 - 7 | 699 | 67 | ||
| AK171266.1 | - | 6 - 7 | 777 | 67 | ||
| AK215485.1 | - | 1 - 2 | 35 | 67 | ||
| BC023432.1 | - | 5 - 6 | 671 | 5029821 | 67 | |
| BC072579.1 | - | 7 - 8 | 1288 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spats2
Genomic Map