Assay Details
| Gene Symbol: Krt75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133357.3 | NP_579935.1 | 4 - 5 | 941 | 83 | |
| GenBank mRNA | AK028972.1 | - | 4 - 5 | 942 | 83 | |
| AK037233.1 | - | 4 - 5 | 942 | 83 | ||
| AK076367.1 | - | 4 - 5 | 942 | 83 | ||
| BC137935.1 | - | 4 - 5 | 942 | 8860954 | 83 | |
| BC137936.1 | - | 4 - 5 | 942 | 8860955 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt75
Genomic Map