Assay Details
| Gene Symbol: Gm30339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035352.1 | - | 1 - 2 | 1561 | 124 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prickle homolog 1 (Drosophila) | Prickle1 |
1110058P22Rik, AW215793, Prickle, b2b019Clo, mpk1 |
-138.1kb | Anti-sense | Chr. 15 - 93499111 - 93596266 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9430014N10 gene | 9430014N10Rik | 168.2kb | Anti-sense | Chr. 15 - 93904856 - 93927111 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm30339
Genomic Map