Assay Details
Gene Symbol: 4933400F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016229.1 | - | 609 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA C030007H22 gene | C030007H22Rik | -244.5kb | Sense | Chr. 1 - 89360342 - 89406133 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
