Assay Details
Gene Symbol: Bcas3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045875.1 | 4 - 5 | 2701 | 151 | ||
GenBank mRNA | AK053124.1 | - | 4 - 5 | 2702 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
