Assay Details
Gene Symbol: Bcas3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045875.1 | 5 - 6 | 2839 | 108 | ||
GenBank mRNA | AK053124.1 | - | 5 - 6 | 2840 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
