Assay Details
| Gene Symbol: Gm34471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041352.1 | - | 4 - 5 | 448 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4921509O07 gene | 4921509O07Rik | -29.2kb | Sense | Chr. 13 - 113446288 - 113502844 on NCBI Build GRCm38 | |
| Downstream | sorting nexin 18 | Snx18 | Snag1 | 57.3kb | Sense | Chr. 13 - 113592179 - 113618564 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm34471
Genomic Map