Assay Details
| Gene Symbol: Ppard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011145.3 | NP_035275.1 | 1 - 2 | 113 | 62 | |
| GenBank mRNA | AK089913.1 | - | 1 - 2 | 61 | 62 | |
| AK154567.1 | - | 1 - 2 | 103 | 62 | ||
| BC070398.1 | - | 1 - 2 | 64 | 5694282 | 62 | |
| CA978861.1 | - | 1 - 2 | 113 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070398.1, NM_011145.3 transcripts.
Set Membership: |
|

Ppard
Genomic Map