Assay Details
| Gene Symbol: Ppard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011145.3 | NP_035275.1 | 2 - 3 | 197 | 69 | |
| GenBank mRNA | AK089913.1 | - | 2 - 3 | 145 | 69 | |
| AK154567.1 | - | 2 - 3 | 187 | 69 | ||
| BC070398.1 | - | 2 - 3 | 148 | 5694282 | 69 | |
| CA978861.1 | - | 2 - 3 | 197 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070398.1, NM_011145.3 transcripts.
Set Membership: |
|

Ppard
Genomic Map